VWR® Traceable® Ultra, Digital Dial Thermometers
Supplier: VWR Collection
61220-417
VWRI620-1665EA 159 SGD
VWRI620-1665 VWRU61220-417
VWR® Traceable® Ultra, Digital Dial Thermometers
Thermometers Digital Thermometers
Dial thermometer for use in soil, cuvettes, test tubes, food products, incubators, biomedical reagents and petroleum hydrocarbons.
  • Easy to read
  • Large, bright LCD
Stainless steel probe (Lר: 120×4,8 mm).
Certifications: Calibration complies with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Delivery information: Supplied with protective case, silver-oxide battery and certificate of calibration.