VWR® Traceable® Alarm Timer, Three Channels
Supplier: VWR Collection
62344-600
VWRU62344-600EA 175 SGD
VWRU62344-600
VWR® Traceable® Alarm Timer, Three Channels
Timers Digital Timers
This timer displays three different times simultaneously, which enables users to run three different tests at the same time. Four rubber feet keep the timer off a wet bench. Perfect unit to dedicate to repetitive tests.
  • Timing capacity: 99 hours, 59 minutes, 59 seconds; quartz-crystal accuracy is 0,001%
  • Functions: Count up/down, time in/out, memories, alarm, clock, adjustable alarm duration/volume
  • Channels may be started individually or simultaneously
  • Timer can concurrently signal the duration of incubating, heating, and cooling
  • When zero is reached, an
More Product Information