VWR® Temperature and Humidity Data Loggers
Supplier: VWR Collection
62344-734
VWRU62344-734EA 170 SGD
VWRU62344-734
VWR® Temperature and Humidity Data Loggers
Data Loggers
This logger is specially designed for use in incubators, store rooms, drying chambers, hoods, cleanrooms, and environmental cabinets.
  • Memory recalls highest and lowest temperature and humidity readings
  • Clock displays AM/PM or 24-hour military time to the exact minute (±0,01%)
  • Unit simultaneously displays ambient temperature, humidity, and time of day
Certifications: Traceable® certificate.
Delivery information: Supplied with stand, wall mount, double-back tape, batteries.