VWR®Traceable®, Digital stopwatch, three-button
Supplier: VWR Collection
62379-234
VWRU62379-234EA 32.2 SGD
VWRU62379-234
VWR®Traceable®, Digital stopwatch, three-button
Timers Digital Stopwatches
This lightweight, inexpensive stopwatch is appropriate for easy-duty, occasional-use needs where high precision is not a requirement.
  • Timing capacity: 23 hours, 59 minutes, 59 seconds (with a resolution of 1/100 of a second for 30 minutes and 1 second thereafter)
  • Functions: Start/stop, time-out, cumulative split, and continuous roll-over timing
  • Also displays time and date
  • Provides a sound confirmation each time a button is pressed
Weight: 24,8 g
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratoryMore Product Information