VWR®, Wash Bottles, Wide Neck, with Coloured Caps, Labeled, Safety-Vented
Supplier: VWR Collection
66001-155 66001-141
VWRU66001-155EA 84.4 SGD
VWRU66001-155 VWRU66001-141
VWR®, Wash Bottles, Wide Neck, with Coloured Caps, Labeled, Safety-Vented
Bottles Wash Bottles
LDPE, with PP spary cap.
  • Unique, 2-piece valve prevents pressure build-up of solvents within these Right-to-Know safety-labelled wash bottles
  • Allows dispensing with bottle upright or inverted
  • Volatiles are vented to eliminate solvent drips
  • Pre-printed with the chemical name and formula, the National Fire Protection Association (NFPA) Section 704 four-colour diamond, health hazards, Chemical Abstract System (CAS) number and suggested protective clothing and equipment
Labelled with six of the most commonly used solvent names or label as per the need.More Product Information