VWR®, Scintillation Vials, with Screw Caps
Supplier: VWR Collection
66022-128 66022-004 66022-060 66022-081 66022-065 66022-106
VWRU66022-128EA 396 SGD
VWRU66022-128 VWRU66022-004 VWRU66022-060 VWRU66022-081 VWRU66022-065 VWRU66022-106
VWR®, Scintillation Vials, with Screw Caps
Vials
These scintillation vials made from borosilicate glass fit all standard counting equipment for liquid scintillation systems.
  • Choose attached or unattached caps
Packaging: Vials are packed in cellular trays, with 100 vials per pray. Trays feature an alphanumeric grid for easy sample identification.