VWR® Solvent-Saver™ , Scintillation vials
Supplier: VWR Collection
66022-300
VWRU66022-300EA 457 SGD
VWRU66022-300
VWR® Solvent-Saver™ , Scintillation vials
Vials
Scintillation vials made from borosilicate glass. The small vial dimensions permit substantial reduction in the quantity of solvent required with resultant savings in solvent costs. The white urea screw cap has a cork-backed foil liner and the top of the cap is suitable for marking.
Packaging: Vials are clean-packed in trays of 200, with 5 trays per case. Caps are clean-packed in plastic bags of 200 pieces.