VWR® Polyclean algaecide Water bath protective agent
Supplier: VWR Collection
71002-500
VWRU71002-500EA 204 SGD
VWRU71002-500
VWR® Polyclean algaecide Water bath protective agent
Cleaning Agents Water Bath Protection Reagents
Prevents growth of algae, keeps baths clean and odour-free.
Bottle of 237 ml solution treats approximately 757 litres of water.
Suitable for use with VWR® refrigerated circulating baths, VWR® heated circulating baths and VWR® circulating open bath systems.