VWR® Spinbar®, Magnetic stirring bars, polygonal
Supplier: VWR Collection
58948-986 58948-988 74950-300 58948-072 58948-080 74950-284 58948-984
VWRU58948-986EA 31.2 SGD
VWRU58948-986 VWRU58948-988 VWRU74950-300 VWRU58948-072 VWRU58948-080 VWRU74950-284 VWRU58948-984
VWR® Spinbar®, Magnetic stirring bars, polygonal
Stirrers Magnetic Stirring Bars
PTFE covered with strong Alnico V magnetic core. The multifaceted surfaces add turbulence relative to smooth cylindrical bars of a similar size.
  • Autoclave safe
  • Pivot ring minimises the contact area between the bar and the vessel
Order Now
SPECIFICATIONS
Environmentally Preferable