VWR®, Variable Speed Rocker
Supplier: VWR Collection
75832-308
VWRU75832-308EA 2400 SGD
VWRU75832-308
VWR®, Variable Speed Rocker
Shakers Rotating/Rocking Shakers
Designed to provide precise speed and tilt angles for a broad range of molecular and biological mixing applications.
  • Variable speed control up to 32 min⁻¹
  • Easily adjustable tilt angle, from 0 to 30°
  • Large 355,6×304,8 mm work surface incudes non-skid rubber pad
  • Suitable for use in the cold room
With variable speed control and adjustable tilt angles, the VWR® Rocker is ideal for many different applications including, gel staining, washing, antibody staining, hybridizations, more.
A large platform is included with a non-skid surface, ideal forMore Product Information