VWR®Standard, Dry block heater, 230 v (export only)
Supplier: VWR Collection
75838-292
VWRU75838-292EA 1910 SGD
VWRU75838-292
VWR®Standard, Dry block heater, 230 v (export only)
Block Heaters Analogue Block Heaters
Multi-purpose, high wattage, constant temperature VWR® Standard Dry Block heaters are economical, versatile and compact.
  • New design with smaller footprint
  • Designed for multi-purpose use
  • Exceptional temperature uniformity
  • Holds interchangeable modular heating blocks
  • Heats to 150 °C (302 °F)
  • Electrical: 230 V, 50/60 Hz, 210 W
Multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a variety of other laboratory procedures.
Each model accepts separate interchangeable modularMore Product Information