VWR®, 96-Well Deep Well Microplates, Extractable-Free
Supplier: VWR Collection
75870-796
VWRU75870-796EA 726 SGD
VWRU75870-796
VWR®, 96-Well Deep Well Microplates, Extractable-Free
Microplates Deep Well Microplates
A range of high quality, dependable deep-well blocks for storage and collection, LC-MS, SPE, and mixing.
  • Guaranteed extractable-free
  • Ready-to-use
  • For assured assays
  • DNase-free and RNase-free
These microplates are made of polypropylene that will not leach unwanted compounds from the polymer.
Plates can be sealed with friction or heat seals to prevent evaporation, condensation, and contamination. These plates are ideal for sample storage and transport. Blocks are rigid and robust, and they can be easily stacked, handled, manipulated, heat sealed,More Product Information