VWR® Measuring Cylinder, Hexagonal Base, TC, with Spout, Class A, Serialised
Supplier: VWR Collection
76004-042
VWRU76004-042EA 97.5 SGD
VWRU76004-042
VWR® Measuring Cylinder, Hexagonal Base, TC, with Spout, Class A, Serialised
Cylinders Measuring Cylinders
VWR® Measuring cylinders are manufactured from ASTM E 438, type 1, class A borosilicate 3.3 glass.
  • Class A serialised measuring cylinders with a hexagonal base
  • Calibrated 'to Contain'
  • Cylinders are made to meet ASTM E 1272 norms
  • Printed double scale
VWR® Measuring cylinders are used for the precise measurement liquid volumes. Cylinders have a large hexagonal base to increase stability. Calibration and tolerances are in compliance to standards: ASTM E1272, ASTM E542, and ASTM E694.
Certifications: Cylinders are supplied with individual work certificates.