VWR® Measuring Cylinders, PP, Class B
Supplier: VWR Collection
76019-256
VWRU76019-256EA 11.9 SGD
VWRU76019-256
VWR® Measuring Cylinders, PP, Class B
Cylinders Measuring Cylinders
These VWR® Measuring cylinders are manufactured from translucent polypropylene.
  • The cylinders feature a stable hexagonal base and a pouring spout
  • All cylinders also feature molded graduations for improved accuracy and consistency
When used continuously, the cylinders have a working temperature range of 0 to 121 °C (32 to 248 °F).
Certifications: Products are Class 'B' as per DIN 12681/ISO 6706. Resin meets US FDA 21 CFR requirements.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable