VWR®, High performance Single Channel Pipettor
Supplier: VWR Collection
76169-234
VWRU76169-234EA 524 SGD
VWRU76169-234
VWR®, High performance Single Channel Pipettor
Pipettes Micropipettes
VWR® High Performance Pipettor is the newest lightweight pipettor designed for user comfort throughout daily work. Thanks to low pipetting forces, users get a unique pipetting experience. With its, high accuracy and precision, the VWR® High Performance Pipettor delivers highly precise and reproducible results.
  • Exclusive volume lock mechanism
  • Lightweight design
  • Low pipetting forces
  • Outstanding precision and accuracy
  • Fully autoclavable without disassembly (120 °C/0,1 MPa/20 min.) and UV resistant
  • Universal tip fitting
  • Unique serial number