VWR® 96-Well Deep Well Plate, Polypropylene
Supplier: VWR Collection
76210-522
VWRU76210-522EA 507 SGD
VWRU76210-522
VWR® 96-Well Deep Well Plate, Polypropylene
Microplates Deep Well Microplates
VWR® 96-well deep well plates are ideal for sample storage, sample re-suspension, cell culture, assay setup, diluting, and compound libraries. They are compatible with automated systems and are widely used in laboratories. Different sealing options are available, including cap seals or film seals, which can be applied independently to avoid cross-contamination.
  • DNase-free, RNase-free, and non-pyrogenic
  • Free from BSE and TSE
  • Latex-free
  • Sterilised by gamma irradiation to SAL 10⁻⁶ (ISO 11137)
  • Three years shelf life following the month of production