VWR® Moisture Analysers
Supplier: VWR Collection
76289-772 76289-774 76290-332 76290-334
VWRU76289-772EA 11010 SGD
VWRU76289-772 VWRU76289-774 VWRU76290-332 VWRU76290-334
VWR® Moisture Analysers
Analysers Moisture Analysers
The VWR® series Moisture analysers offer precise and reliable results for moisture content analysis.
  • Automatic opening and closing of drying chamber lid
  • Large, 5" colour capacitive touch screen
  • Moisture content analysis
  • Dry mass determination
  • Hands-free operation with IR proximity sensors
  • Three-year warranty
The VWR® Moisture analysers can be utilized for a range of applications. Choose between an IR Emitter (max temperature 160 °C) or Halogen (max temperature 250 °C) heating module. For accurate moisture content analysis, the VWR® moisture analyserMore Product Information