VWR®, Digital General-Purpose Water Bath
Supplier: VWR Collection
76308-898 76308-896 76308-906 76308-830 76308-832 76308-834 76308-900 76308-902 76308-904
VWRU76308-898EA 2110 SGD
VWRU76308-898 VWRU76308-896 VWRU76308-906 VWRU76308-830 VWRU76308-832 VWRU76308-834 VWRU76308-900 VWRU76308-902 VWRU76308-904
VWR®, Digital General-Purpose Water Bath
Baths Water Baths, Unstirred
These premium digital general-purpose water baths are designed with high-gabled lids with hinge to hold lid up and allow condensate to drain into bath.
  • Easy-to-read, colour, touch screen
  • Three push-button pre-sets for frequently used temperatures
  • Large buttons for easy navigation
  • Eco mode for heat-off use
  • Temperature calibration capability
  • Safety thermostat and alarms
Features five language (Chinese, English, French, German and Spanish) setting option and have rear-mounted drain on 10, 20 and 18 L models.
Temperature stability: ±0,2 °C
Certifications: cETLus compliant.
Order Now
SPECIFICATIONS
Environmentally Preferable