VWR® Sharp Container with Flip Up Lid
Supplier: VWR Collection
76319-860
VWRU76319-860EA 5.1 SGD
VWRU76319-860
VWR® Sharp Container with Flip Up Lid
Bins Sharps Containers
Container provide for the safe disposal of sharps and biohazard waste. The flip up lid design is perfect for busy professional in settings such as laboratories, hospitals, dental, or physician offices requiring one-handed access to open and close sharps container.
  • Flip Up lid is intuitive to use, allowing one-handed operation
  • Designed for use in high activity environments
  • Translucent color with lead-free pigments containing no heavy metals
  • Constructed of puncture and impact resistant material
  • Safe to autoclave or incinerate
  • Stackable design
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable