VWR®, Universal pipette tips
Supplier: VWR Collection
76322-144
VWRU76322-144EA 7.8 SGD
VWRU76322-144
VWR®, Universal pipette tips
Pipette Tips
VWR® Universal low retention pipette tips repel liquid by creating a hydrophobic surface on the inside of the tip increasing your accuracy and data quality.
  • Compatible with the widest range of pipette
  • Maximizes sample recovery with low binding resin and a fine point design
  • Improves ergonomics by reducing the force required to attach and eject a tip
  • Autoclavable at 1 bar, 122 °C for 15 minutes
  • Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to ten times more hydrophobic than other polypropyleneMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable