VWR® DuraChill® Recirculating Chillers, –10 to 70 °C, 240 V
Supplier: VWR Collection
76472-554
VWRU76472-554EA 15880 SGD
VWRU76472-554
VWR® DuraChill® Recirculating Chillers, –10 to 70 °C, 240 V
Circulating Thermostats Closed Circulator Systems Cooling Circulators
DuraChill® chillers provide stable and reliable cooling for various common heat removal applications, including laser etching, AA furnaces, ICP, rotary evaporators, vacuum systems, reaction vessels, plasma etching, and condenser cooling.
  • Large, full-color touchscreen display with intuitive interface and support for five different languages
  • Compact, portable design takes up less floor space
  • DynamicFilter™ self-changing filter system
  • Front fill reservoir
  • Liquid level monitoring to prevent pump failure
  • Ultraviolet anti-biological light system
  • User-adjustable
More Product Information