VWR® Sterile Standard Square Weighing Boats, White
Supplier: VWR Collection
76472-584
VWRU76472-584EA 1400 SGD
VWRU76472-584
VWR® Sterile Standard Square Weighing Boats, White
Boats Weighing Boats
These square weighing boats are flexible and feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
  • Electron beam sterilised to reduce the risk of cross-contamination and compromised samples
  • Made from 100% virgin anti-static polystyrene that is waterproof and reduces the chance of materials sticking to the units
  • Disposable
  • Can be used with microwave moisture analyzers
  • Rounded corners simplify transfer and allow flexing for controlled pouring of liquid, powdered, or granular samples
  • Boats are shallow with a wide, flat bottom to
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable