VWR® Extended Cuff Nitrile Gloves
Supplier: VWR Collection
76582-342
VWRU76582-342EA 19.7 SGD
VWRU76582-342
VWR® Extended Cuff Nitrile Gloves
Gloves
Ambidextrous gloves with extended cuff are extra-soft and comfortable for everyday use. They protect hands from a broad range of chemicals.
  • 12" extended cuff
  • Nonsterile
  • Latex-free
  • Extra-soft and comfortable
  • Powder-free
  • Beaded cuff allows easy donning
Ideal for use in industrial and laboratory applications. Made of premium compounded nitrile for high manufacturing quality measures and product consistency. Gloves protect against biohazards and a broad range of chemicals. Composition is 100% latex-free to eliminate adverse reactions associated withMore Product Information