VWR®, Spatulas, Disposable
Supplier: VWR Collection
80081-190 80081-194 80081-192 80081-188 89097-816
VWRU80081-190EA 83.7 SGD
VWRU80081-190 VWRU80081-194 VWRU80081-192 VWRU80081-188 VWRU89097-816
VWR®, Spatulas, Disposable
Spatulas
PP. Multipurpose, robust and inexpensive tools for collecting, transferring and processing samples of chemical and biological materials.
  • Resistant to dilute acids and alkalis
  • Reduce cross-contamination
  • Autoclavable
  • Compliant with FDA standards for applications involving contact with food
The innovative shape of these spatulas allows them to be used as a spatula, spoon, scoop, scraper, mixing rod or for wiping. Ideal for recovering samples that have been frozen in liquid nitrogen. Standard spatulas feature a unique spatula/spoon end and a smaller scoopMore Product Information