VWR®, Write-On Label Rolls, Magnetic
Supplier: VWR Collection
80094-278
VWRU80094-278EA 220 SGD
VWRU80094-278
VWR®, Write-On Label Rolls, Magnetic
Labels
This magnetic label material has a white vinyl surface that may be written on with wet-erase marker and then wiped off. Provided in a roll that may be cut to any size strip.
  • Thickness: 0,8 mm