VWR®, Laboratory Notebooks
Supplier: VWR Collection
82007-219 82026-407 82021-133 82021-131 82021-135
VWRU82007-219EA 45.2 SGD
VWRU82007-219 VWRU82026-407 VWRU82021-133 VWRU82021-131 VWRU82021-135
VWR®, Laboratory Notebooks
Notebooks
The notebook covers are waterproof and chemical-resistant. They also contain periodic and conversion tables in the back.
  • Acid-free
  • Grid or lined
  • Place for signature on the bottom of every page
L×W: 281×216 mm