VWR®, Disposal Box for Pipette Tips
Supplier: VWR Collection
82010-754
VWRU82010-754EA 325 SGD
VWRU82010-754
VWR®, Disposal Box for Pipette Tips
Boxes Beta/Gamma Radiation Boxes
Acrylic, clear.
  • Eliminates any need to handle pipette tips used with hazardous fluids
  • The 10 mm thick acrylic provides protection from beta radiation
  • Large, hinged lid allows easy removal of contents
This box features a flip-top port cover for convenient one-handed disposal of used pipette tips.