VWR®, Microtube and Test Tube Racks, Floating
Supplier: VWR Collection
82017-630
VWRU82017-630EA 35 SGD
VWRU82017-630
VWR®, Microtube and Test Tube Racks, Floating
Racks Tube Racks
Polyfoam, coloured.
  • These stands float and remain on the surface of the bath, preventing the vessels they contain from floating free
  • A variety of geometrical shapes enable individual and easily sortable arrangement in the water bath
  • With handles for easy insertion and removal
For incubating sample, centrifuge and microcentrifuge tubes in water baths.