VWR® Racks for Centrifuge Tubes
Supplier: VWR Collection
82024-452
VWRU82024-452EA 76.7 SGD
VWRU82024-452
VWR® Racks for Centrifuge Tubes
Racks Tube Racks
Steel with an epoxy coating, autoclavable.
  • Accepts tubes up to 30 mm diameter
  • Ribbed base for better stability of the tubes
  • Very good chemical resistance
For 50 ml centrifuge tubes. Can also be used for all other types of tubes.
Order Now
SPECIFICATIONS
Environmentally Preferable