VWR®, Microtube Racks, 80 Place
Supplier: VWR Collection
82024-479
VWRU82024-479EA 44.1 SGD
VWRU82024-479
VWR®, Microtube Racks, 80 Place
Racks Tube Racks
Fraction collector plate style rack made of PP.
  • Moulded-in coordinates to aid orientation
  • Conical shaped holes (Ø 11 mm) to hold microtubes securely
  • Easily fill and transfer 80 tubes in compact arrangement (5×16 array)