VWR®, Spatulas
Supplier: VWR Collection
82027-462
VWRU82027-462EA 30 SGD
VWRU82027-462
VWR®, Spatulas
Spatulas
Stainless steel spatulas with wooden handle. Economical tools for handling, transferring, blending, or collecting chemical and biological samples.
  • Sturdy and durable
  • Stainless steel construction
  • Multiple sizes allow for varied laboratory applications
Order Now
SPECIFICATIONS
Environmentally Preferable