VWR®, Spatula, PTFE Coated
Supplier: VWR Collection
82027-476
VWRU82027-476EA 42.2 SGD
VWRU82027-476
VWR®, Spatula, PTFE Coated
Spatulas
Stainless steel, PTFE coated, with wooden handle.
Sturdy, economical tools for handling, transferring, blending, or collecting chemical and biological samples. They can be used to transfer sticky or corrosive materials without scratching or damaging container surfaces.