VWR®, Double Spatulas
Supplier: VWR Collection
82027-492 82027-496 82027-494 82027-528
VWRU82027-492EA 40.1 SGD
VWRU82027-492 VWRU82027-496 VWRU82027-494 VWRU82027-528
VWR®, Double Spatulas
Spatulas
Stainless steel. These double spatulas with one rounded and one flat end are ideal for transferring, collecting, mixing, or handling chemical or biological samples.
  • Versatile design enables use as a spatula, spoon, scraper, or stirring rod
  • Spatulas feature one rounded end and one flat end
  • Non stick surfaces
  • Spatula 82027-494 (305 mm length) features heavy-duty construction
Order Now
SPECIFICATIONS
Environmentally Preferable