VWR®, Spatulas, Stainless Steel/PVC Handle
Supplier: VWR Collection
82027-506 82027-502
VWRU82027-506EA 21.8 SGD
VWRU82027-506 VWRU82027-502
VWR®, Spatulas, Stainless Steel/PVC Handle
Spatulas
Stainless steel, PVC handle. Sturdy, economical tools for handling, transferring, blending, or collecting chemical and biological samples.
  • PVC handle is resistant to acids and most solvents
  • Handle can withstand boiling water
  • Multiple sizes allow for varied laboratory applications
Order Now
SPECIFICATIONS
Environmentally Preferable