VWR®, Micro Spatulas
Supplier: VWR Collection
82027-518 82027-520
VWRU82027-518EA 16.4 SGD
VWRU82027-518 VWRU82027-520
VWR®, Micro Spatulas
Spatulas
Stainless steel. These micro spatulas are ideal for small quantity applications such as sample transfer into test tubes or microcentrifuge tubes.
  • Rustproof material
  • With non stick surfaces
  • Mirror finish
They feature one rounded end and one tapered end, allowing for a variety of transfer, mixing, or handling applications.
Order Now
SPECIFICATIONS
Environmentally Preferable