VWR®, Double Spatulas, Hayman Style
Supplier: VWR Collection
82027-522 82027-524
VWRU82027-522EA 163 SGD
VWRU82027-522 VWRU82027-524
VWR®, Double Spatulas, Hayman Style
Spatulas
Stainless steel. These spatulas are ideal for applications involving the transfer or mixing of small sample amounts.
Model 82027-522 features a round spatula on one end and a spoon on the other. Model 82027-524 features a tapered, bent-tip spatula on one end and a spoon on the other. Efficient design reduces the risk of sample loss.
Order Now
SPECIFICATIONS
Environmentally Preferable