VWR®, Spoon Spatula, Stainless Steel
Supplier: VWR Collection
82027-532
VWRU82027-532EA 15.2 SGD
VWRU82027-532
VWR®, Spoon Spatula, Stainless Steel
Spatulas
Stainless steel. This double spatula is ideal for use in the transfer, collection, and handling of chemical or biological samples.
  • Rustproof stainless steel
  • Flat spatula at one end and a spoon at the other
  • Substances will not stick to spatula surfaces