VWR®, Spatula
Supplier: VWR Collection
82027-534
VWRU82027-534EA 26.9 SGD
VWRU82027-534
VWR®, Spatula
Spatulas
Stainless steel, PTFE coated. This spatula features one rounded end and one flat end, with PTFE coating throughout.
Can be used to transfer corrosive or sticky materials without scratching or damaging container surfaces.