VWR® Oil-only, Absorbent mat pads and rolls
Supplier: VWR Collection
82028-020 82028-022
VWRU82028-020EA 227 SGD
VWRU82028-020 VWRU82028-022
VWR® Oil-only, Absorbent mat pads and rolls
Spill Control Materials
Absorbent pads and rolls made from PP are ideal for spill cleanup; fast-wicking dimpled design absorbs liquids quickly to leave area clean and dry for improved safety. Efficient high-absorbency materials reduce waste of absorbents. Oil-only pads and rolls absorb oils but repel water.
  • Soak up leaks and spills fast, keeping floors at your workplace clean, dry and safe
  • High-capacity absorbents stay on the job longer and are perfect for stocking spill kits
  • Can be incinerated after use to reduce waste