VWR®, PCR Plates, 96 Well
Supplier: VWR Collection
83007-374
VWRU83007-374EA 71.3 SGD
VWRU83007-374
VWR®, PCR Plates, 96 Well
Microplates PCR Plates
PP. These PCR plates are compatible with most thermal cyclers, including the Applied Biosystems 9600 and 9700, and the MJ Research PTC-100, and are ideal for high throughput screening thermal cycler applications.
  • Smooth, thin, uniform well walls ensure accurate thermal transfer
  • Plates are thin, flexible and easy to cut
  • Certified free from DNase, RNase and human genomic DNA
  • Printed alphanumeric labelling and cut corner simplifies plate orientation and sample identification
Working capacity: 200 µl