VWR®, Laboratory Notebooks
Supplier: VWR Collection
89005-138 89005-136 89005-132
VWRI818-0074EA 138 SGD
VWRI818-0074 VWRI818-0075 VWRI818-0076 VWRI818-0077 VWRI818-0078 VWRI818-0079 VWRI818-0080 VWRI818-0081 VWRI818-0082 VWRI818-0083 VWRI818-0084 VWRI818-0086 VWRI818-0087 VWRI818-0088 VWRI818-0089 VWRI818-0090 VWRI818-0091 VWRI818-0092 VWRI818-0093 VWRI818-0094 VWRI818-0095 VWRI818-0096 VWRI818-0097 VWRI818-0098 VWRI818-0099 VWRI818-0068 VWRI818-0069 VWRI818-0070 VWRI818-0071 VWRI818-0072 VWRI818-0073 VWRI818-0112 VWRI818-0113 VWRI818-0114 VWRI818-0115 VWRU89005-138 VWRU89005-136 VWRU89005-132 VWRI818-0282 VWRI818-0283
VWR®, Laboratory Notebooks
Notebooks
Imitation leather cover with coating, for durability in laboratory environments. Acid-free, archive safe paper (90 gsm). Includes laboratory notebook guidelines page, table of contents and user data page. Available in different language formats: English only, English-French, English-German, English-Spanish and English-Italian.
  • Pages are sewn in sections with round binding so that book lays flat
  • Scientific 5 mm grid or ruled format with date and witness signature blocks
  • Waterproof
Format: A4 (21×29,7 cm) or A5 (14,8×21 cm). A4 books available in threeMore Product Information