VWR® UV Guard®, Storage bags
Supplier: VWR Collection
89005-326 89005-328 89005-332 89005-324 89005-320 89005-318
VWRU89005-326EA 207 SGD
VWRU89005-326 VWRU89005-328 VWRU89005-332 VWRU89005-324 VWRU89005-320 VWRU89005-318
VWR® UV Guard®, Storage bags
Bags Storage Bags
UV Guard® UV protection bag is the amber UV blocker protective bags with the Color Line™. It prevents the bag contents from deterioration.
  • Premium zipper opens quickly and easily
  • Amber color makes identification of light-sensitive contents easy
Complies with USP XXII and USP XXIII procedure 66,1 with light transmission not exceeding 10% at any wavelength from 290 to 450 nm.
Order Now
SPECIFICATIONS
Environmentally Preferable