VWR®, Glove box holder
Supplier: VWR Collection
89011-724
VWRU89011-724EA 168 SGD
VWRU89011-724
VWR®, Glove box holder
Glove Dispensers and Glove Box Holders
These acrylic holders are constructed from tough, clear acrylic so that you can clearly see the size and material of the gloves within. Each holder includes a polyester foam insert to securely hold a range of glove box sizes.
  • Models available which hold one, two, or three glove boxes
Delivery information: Wall-mount screws included.