VWR®, Sample/centrifuge tubes, with plug seal cap
Supplier: VWR Collection
89012-780
VWRU89012-780EA 233 SGD
VWRU89012-780
VWR®, Sample/centrifuge tubes, with plug seal cap
Tubes Centrifuge Tubes
PP, clear, with translucent PP plug seal cap.
These tubes features moulded graduations and a leakproof plug seal cap designed to withstand 95 kPa of pressure. Skirted design enables tubes to stand upright, allowing for use without separate racks or stands.
Packaging: Bulk packaged with 500 separate caps.