VWR®, Tissue Grinders, PTFE
Supplier: VWR Collection
89026-386 89026-398 89026-392 89026-404
VWRU89026-386EA 198 SGD
VWRU89026-386 VWRU89026-398 VWRU89026-392 VWRU89026-404
VWR®, Tissue Grinders, PTFE
Homogenisers Tissue Grinders
PTFE, stainless steel and precision borosilicate glass. Designed to control the reduction of particle size and homogenisation of variety of substances (biological material). Shearing forces will be generated by the movement of a rotating plunger in a precision bore tube.
  • Chemically inert and leak free
  • Non-contaminating
  • Usable up to 280 °C
Certifications: Approved by FDA and USP Class VI.
Order Now
SPECIFICATIONS
Environmentally Preferable