VWR®, Sleeves for Ground Joints, PTFE
Supplier: VWR Collection
89026-490
VWRU89026-490EA 97.6 SGD
VWRU89026-490
VWR®, Sleeves for Ground Joints, PTFE
NS Joint Sleeves and Connectors Joint Sleeves
These PTFE sleeves withstand low to medium pressure and are inserted between standard ground glass joints to eliminate the use of grease and prevent sticking.
  • Chemically resistant
  • Inert
  • Usable upto 280 °C (536 °F)
Sleeves are inert, have a wall thickness of about 0,05 mm.