VWR®, Critical Swab with Cone-Shaped Reticulated Foam Head
Supplier: VWR Collection
89031-282
VWRU89031-282EA 237 SGD
VWRU89031-282
VWR®, Critical Swab with Cone-Shaped Reticulated Foam Head
Swabs Swabs for Sampling
Swab features reticulated foam and a blue glass-filled polypropylene shaft for extra rigidity.
For use in critical environments.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904