VWR®, Bitumen Filter Crucible
Supplier: VWR Collection
89038-036
VWRU89038-036EA 23.4 SGD
VWRU89038-036
VWR®, Bitumen Filter Crucible
Crucibles
Porcelain, with perforated bottom, glased inside and out, except base and rim. Ideal for total suspended solids determination. For use with 30 to 33 mm diameter filter paper.
Order Now
SPECIFICATIONS
Environmentally Preferable