VWR®, Gooch Crucibles
Supplier: VWR Collection
89038-042
VWRU89038-042EA 124 SGD
VWRU89038-042
VWR®, Gooch Crucibles
Crucibles
Porcelain gooch crucibles, with perforated bottom, glazed inside and out, except base and rim. Ideal for total suspended solids determination. Crucible 89038-046 has two holes for suspending in extraction apparatus.
  • For use in temperatures up to 1150 °C
Ordering information: Lids need to be ordered seperately.
Order Now
SPECIFICATIONS
Environmentally Preferable