VWR®, Rose crucibles, Porcelain
Supplier: VWR Collection
89038-052
VWRU89038-052EA 121 SGD
VWRU89038-052
VWR®, Rose crucibles, Porcelain
Crucibles
Porcelain, unglazed crucibles.
  • Withstand maximum temperature of 1150 °C